Learn More
Turbidity 0.5 NTU Calibration Standard, Formazin Certified Reference Material, MilliporeSigma™ Supelco™
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
$150.46 - $346.39
Specifications
Grade Certified Reference Material
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111001787
View Documents
Merck Emd Millipore
TURB05100ML
100 mL
Each for $150.46
Only null left
111001776
View Documents
Merck Emd Millipore
TURB05500ML
500 mL
Each for $211.84
Only null left
111001769
View Documents
Merck Emd Millipore
TURB051L
1 L
Each for $346.39
Only null left
Description
Description
Specifications
Specifications
Certified Reference Material