missing translation for 'onlineSavingsMsg'
Learn More

Tungsten wire basket; # turns, 8; ID(mm), 7; Ht(mm), 9; Wire leads(cm), 5.0; Wire dia(mm), 0.51, Thermo Scientific™

Catalog No. AA41178KK
Change view
Click to view available options
Quantity:
10 Pc.
50 Pc.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41178KK 10 Pc.
AA41178KD 50 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA41178KK Supplier Thermo Scientific Chemicals Supplier No. 41178KK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Quantity 10 Pc.
Form Wire
Chemical Name or Material Tungsten wire basket, 8 turns, ID(mm), 7; Ht(mm), 9; Wire leads(cm), 5.0; Wire dia(mm), 0.51
EINECSNumber 231-143-9
TSCA Yes