missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungsten wire, 0.015mm (0.00059in) dia, 99.95% (metals basis)
Form: approx. 0.003g/m | CAS: 7440-33-7 | W | 183.84 g/mol
$118.99 - $404.26
Chemical Identifiers
| CAS | 7440-33-7 |
|---|---|
| Molecular Formula | W |
| Molecular Weight (g/mol) | 183.84 |
| MDL Number | MFCD00011461 |
| InChI Key | WFKWXMTUELFFGS-UHFFFAOYSA-N |
| Synonym | wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder |
| PubChem CID | 23964 |
| ChEBI | CHEBI:27998 |
| IUPAC Name | tungsten |
| SMILES | [W] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-33-7 | |
| 183.84 | |
| WFKWXMTUELFFGS-UHFFFAOYSA-N | |
| 23964 | |
| tungsten |
| W | |
| MFCD00011461 | |
| wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder | |
| CHEBI:27998 | |
| [W] |
Specifications
| Silver to Gray | |
| Silver Gray wirw | |
| 25 m | |
| Wire | |
| MFCD00011461 | |
| WFKWXMTUELFFGS-UHFFFAOYSA-N | |
| tungsten | |
| 23964 | |
| Tungsten |
| 7440-33-7 | |
| 19.3g/cm3 | |
| 99.95% (Metals basis) | |
| W | |
| wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder | |
| [W] | |
| 183.84 | |
| CHEBI:27998 |
Safety and Handling
EINECSNumber : 231-143-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only