missing translation for 'onlineSavingsMsg'
Learn More

Tungsten wire, 1.6mm (0.063in) dia, annealed, 99.95% (metals basis)

Catalog No. AA43312G1
Change view
Click to view available options
Quantity:
1 m
5 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43312G1 1 m
AA43312G5 5 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43312G1 Supplier Thermo Scientific Chemicals Supplier No. 043312G1

CAS: 7440-33-7 | W | 183.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Quantity 1 m
Weight ≈38.8g/m
Form Wire
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Tungsten wire, 1.6mm (0.063 in.) dia., Annealed, Temper: soft
EINECSNumber 231-143-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only