missing translation for 'onlineSavingsMsg'
Learn More

Tungsten wire, 0.5mm (0.02in) dia, annealed, 99.95% (metals basis)

Catalog No. p-5547786
Change view
Click to view available options
Quantity:
5 m
25 m
100 m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10409G5 5 m
AA10409G9 25 m
AA10409H4 100 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA10409G5 Supplier Thermo Scientific Chemicals Supplier No. 010409G5
Only null left

CAS: 7440-33-7 | W | 183.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Appearance Silver Gray wire
Density 19.3g/cm3
Quantity 5 m
Form Wire
Assay Percent Range 99.95% (Metals basis)
Chemical Name or Material Tungsten
EINECSNumber 231-143-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only