missing translation for 'onlineSavingsMsg'
Learn More

Tungsten sulfide, 99.8% (metals basis)

Catalog No. p-4904565
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1182918 50 g
AA1182930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1182918 Supplier Thermo Scientific Chemicals Supplier No. 01182918
Only null left

CAS: 12138-09-9 | S2W | 247.96 g/mol

Tungsten sulfide is used as a component of catalysts for hydrotreating of crude oil, hydrodesulfurization and hydrodenitrification reactions. It finds application as solid-state secondary lithium battery cathodes and other electrochemical devices. It is found that it forms inorganic nanotubes, which is better reinforcing agents than carbon nanotubes. Tungsten sulfide addition to epoxy resin improves adhesion, fracture toughness and strain energy release rate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12138-09-9
Molecular Weight (g/mol) 247.96
MDL Number MFCD00011467
InChI Key ITRNXVSDJBHYNJ-UHFFFAOYSA-N
IUPAC Name disulfanylidenetungsten
SMILES S=[W]=S

Specifications

Melting Point 1250°C (decomposition)
Color Black
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.8% metal basis
Molecular Formula S2W
Solubility Information Slightly soluble in water.
Formula Weight 247.98
Odor Odorless
Density 7.5
Chemical Name or Material Tungsten sulfide
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 235-243-3
RTECSNumber YO7716000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only