accessibility menu, dialog, popup

UserName

SWS

CAS RN 12138-09-9

IUPAC Name: disulfanylidenetungsten
Synonyms: tungsten disulfide|tungsten disulphide
Molecular Weight (g/mol): 247.96
Molecular Formula: S2W
InChi Key: ITRNXVSDJBHYNJ-UHFFFAOYSA-N
SMILES: S=[W]=S
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results