missing translation for 'onlineSavingsMsg'
Learn More

Tungsten rod, 2mm (0.08in) dia, 99.95% (metals basis)

Catalog No. p-4412912
Change view
Click to view available options
Quantity:
30 cm
2 x 10 cm
5 x 30 cm
Catalog No. Quantity
AA439719X 2 x 10 cm
AA43971BV 30 cm
AA43971CE 5 x 30 cm
3 options

Catalog No. AA439719X

Supplier: Thermo Scientific Chemicals 0439719X

Only null left

Form: approx. 6.1g/10cm | CAS: 7440-33-7 | W | 183.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Color Silver to Gray
Appearance Silver Gray rod
Quantity 2 x 10 cm
Form Rod
Assay Percent Range 99.95% (Metals basis)
Chemical Name or Material Tungsten
EINECSNumber 231-143-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only