missing translation for 'onlineSavingsMsg'
Learn More

Tungsten rod, 3.175mm (0.125in) dia, annealed, 99.95% (metals basis)

Catalog No. p-4782217
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 cm
25 cm
5 x 100 cm
Catalog No. Quantity
AA10407BU 25 cm
AA10407BY 100 cm
AA10407CQ 5 x 100 cm
3 options

Catalog No. AA10407BU

Supplier: Thermo Scientific Chemicals 010407BU

Only null left

Form: Approx. 1.53g/cm; Temper: soft | CAS: 7440-33-7 | W | 183.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Color Silver to Gray
Appearance Silver Gray
Quantity 25 cm
Form Rod
Assay Percent Range 99.95% (Metals basis)
Chemical Name or Material Tungsten, Annealed
EINECSNumber 231-143-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only