missing translation for 'onlineSavingsMsg'
Learn More

Tungsten rod, 12.7mm (0.5in) dia, 99.95% (metals basis)

Catalog No. p-7048396
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 cm
5 cm
Catalog No. Quantity
AA10403BQ 5 cm
AA10403BU 25 cm
2 options

Catalog No. AA10403BQ

Supplier: Thermo Scientific Chemicals 010403BQ

Only null left

CAS: 7440-33-7 | W | 183.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Quantity 5 cm
Weight ≈24.4g/cm
Form Rod
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Tungsten rod, 12.7mm (0.5 in.) dia.
EINECSNumber 231-143-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only