missing translation for 'onlineSavingsMsg'
Learn More

Tungsten rod, 1.5mm (0.06in) dia, 99.95% (metals basis)

Catalog No. AA42233CE
Change view
Click to view available options
Quantity:
30 cm
5 x 30 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42233CE 5 x 30 cm
AA42233BV 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42233CE Supplier Thermo Scientific Chemicals Supplier No. 042233CE

CAS: 7440-33-7 | W | 183.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Quantity 5 x 30 cm
Weight ≈3.4g/10cm
Form Rod
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Tungsten rod, 1.5mm (0.06 in.) dia.
EINECSNumber 231-143-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only