missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungsten powder, APS 4-6 micron, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Gray |
| Quantity | 10 g |
| Particle Size | APS 4 to 6 Micron |
| Form | Powder |
| UN Number | UN3089 |
| Percent Purity | 99.999% |
| Grade | Puratronic |
| Concentration or Composition (by Analyte or Components) | Mo <1ppm |
| Assay | (metals basis) |
| Chemical Name or Material | Tungsten powder |
Safety and Handling
| Hazard Category | H228-H500 |
| Hazard Statement | GHS H Statement H228 Flammable solid. |
| Precautionary Statement | P210-P240-P241-P280-P370+P378q |
| DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S. |
| EINECSNumber | 231-143-9 |
| RTECSNumber | YO7175000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only