missing translation for 'onlineSavingsMsg'
Learn More

Tungsten, plasma standard solution, Specpure™ W 1000μg/mL

Catalog No. AA35770AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35770AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA35770AE Supplier Thermo Scientific Chemicals Supplier No. 035770AE

W in 5% HNO3/tr. HF

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

Molecular Formula W in 5% HNO3/tr. HF
MDL Number MFCD00011461

Specifications

Odor Acidic
Physical Form Liquid
Concentration W 1000 μg/mL
UN Number UN3264
Quantity 100 mL
Chemical Name or Material Tungsten, plasma standard solution, Specpure™
Hazard Category H290-H314-H335
Hazard Statement GHS H Statement
H314-H302-H312-H332;
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only