Learn More
Tungsten, plasma standard solution, Specpure™ W 1000μg/mL
(NH4)2WO4 in H2O
Supplier:  Thermo Scientific Chemicals 042248AP
 View more versions of this product
Catalog No. AA42248AP
Only null left
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Tungsten, plasma standard solution, Specpure™
(NH4)2WO4 in H2O
MFCD00011461
Liquid
W 1000 μg/mL
500 mL
Chemical Identifiers
(NH4)2WO4 in H2O
MFCD00011461
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures