missing translation for 'onlineSavingsMsg'
Learn More

Tungsten ICP Standard, 1000 ppm W in H2O/tr NH4OH, Ricca Chemical

Single Element ICP Standard

$604.03 - $604.03

Chemical Identifiers

CAS 11120-25-5
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PW1KW500
View Documents
Ricca Chemical Company
PW1KW500
500 mL Natural Poly Bottle
Each for $604.03
Only null left
 
Description

Description

Specifications

Chemical Identifiers

11120-25-5

Specifications

Natural Poly Bottle
97.0,0.49,0.14
Liquid
100°C
500 mL
Tungsten ICP Standard
11120-25-5 , 1336-21-6 , 11120-25-5
100.0,0.51,0.14
Colorless
Standard
1mL = 1mg W (1, 000ppm W)(NH4)2WO4 in H2O / tr NH4OH
Videos
Safety and Handling

Safety and Handling

ShelfLife : 18 months

SDS
Documents

Documents