missing translation for 'onlineSavingsMsg'
Learn More

Tungsten gauze, 20 mesh woven from 0.127mm (0.005in) dia wire

Catalog No. AA40922HB
Change view
Click to view available options
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40922HB 300 x 300 mm
AA40922FL 75 x 75 mm
AA40922GT 150 x 150 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA40922HB Supplier Thermo Scientific Chemicals Supplier No. 040922HB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Form: Wire Cloth; Open area: 81%; Width of opening: 0.045 in. | CAS: 7440-33-7 | W | 183.84 g/mol

Filtration, electrodes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Quantity 300 x 300 mm
Mesh Size 20 Mesh
Form Wire Cloth
Chemical Name or Material Tungsten
EINECSNumber 231-143-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only