missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungsten gauze, 100 mesh woven from 0.0509mm (0.002in) dia wire
CAS: 7440-33-7 | W | 183.84 g/mol
Supplier: Thermo Scientific Chemicals 040925GT
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Tungsten gauze | |
| 7440-33-7 | |
| 100 Mesh | |
| W | |
| wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder | |
| [W] | |
| 183.84 | |
| CHEBI:27998 |
| Woven from 0.0509mm (0.002 in.) dia. wire | |
| 150 x 150 mm | |
| Wire Cloth | |
| MFCD00011461 | |
| WFKWXMTUELFFGS-UHFFFAOYSA-N | |
| tungsten | |
| 23964 |
Chemical Identifiers
| 7440-33-7 | |
| 183.84 | |
| WFKWXMTUELFFGS-UHFFFAOYSA-N | |
| 23964 | |
| tungsten |
| W | |
| MFCD00011461 | |
| wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder | |
| CHEBI:27998 | |
| [W] |
Safety and Handling
EINECSNumber : 231-143-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only