missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungsten foil, 0.05mm (0.002in) thick, 99.95% (metals basis)
CAS: 7440-33-7 | W | 183.84 g/mol
$102.22 - $868.53
Chemical Identifiers
| CAS | 7440-33-7 |
|---|---|
| Molecular Formula | W |
| Molecular Weight (g/mol) | 183.84 |
| MDL Number | MFCD00011461 |
| InChI Key | WFKWXMTUELFFGS-UHFFFAOYSA-N |
| Synonym | wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder |
| PubChem CID | 23964 |
| ChEBI | CHEBI:27998 |
| IUPAC Name | tungsten |
| SMILES | [W] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA10417FI
|
Thermo Scientific Chemicals
010417FI |
50 x 50 mm |
Each for $102.22
|
|
|||||
|
AA10417GH
|
Thermo Scientific Chemicals
010417GH |
100 x 100 mm |
Each for $255.97
|
|
|||||
|
AA10417HP
|
Thermo Scientific Chemicals
010417HP |
100 x 500 mm |
Each for $868.53
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-33-7 | |
| 183.84 | |
| WFKWXMTUELFFGS-UHFFFAOYSA-N | |
| 23964 | |
| tungsten |
| W | |
| MFCD00011461 | |
| wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder | |
| CHEBI:27998 | |
| [W] |
Specifications
| 7440-33-7 | |
| ≈2.4g/50x50mm | |
| W | |
| wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder | |
| [W] | |
| 183.84 | |
| CHEBI:27998 | |
| Tungsten foil, 0.05mm (0.002 in.) thick |
| 50 x 50 mm | |
| Foil | |
| MFCD00011461 | |
| WFKWXMTUELFFGS-UHFFFAOYSA-N | |
| tungsten | |
| 23964 | |
| 99.95% | |
| (metals basis) |
Safety and Handling
EINECSNumber : 231-143-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only