missing translation for 'onlineSavingsMsg'
Learn More

Tungsten carbide, 99.5% (metals basis)

CAS: 12070-12-1 | CW | 195.85 g/mol

$187.12 - $656.70

Chemical Identifiers

CAS 12070-12-1
Molecular Formula CW
Molecular Weight (g/mol) 195.85
MDL Number MFCD00011464
InChI Key UONOETXJSWQNOL-UHFFFAOYSA-N
Synonym tungsten carbide, tungsten iv carbide, rfdbds g@, tungsten carbide ds 60, methanidylidynetungstenylium, methanidylidynetungsten 1+, tungsten-carbide/cobalt co=8wt% powder nano
IUPAC Name methanidylidynetungstenylium
SMILES [C-]#[W+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1248222
View Documents
Thermo Scientific Chemicals
01248222
100 g
Each for $187.12
Only null left
 
AA1248236
View Documents
Thermo Scientific Chemicals
01248236
500 g
Each for $656.70
Only null left
 
Description

Description

Tungsten carbide is extensively used as a cutting tool for machining and in armor-piercing ammunition. It is also used in farriery, the shoeing of horses as well as to improve traction on slippery surfaces like roads or ice. It finds application in nuclear reactors as an effective neutron reflector. It is also used for making surgical instruments meant for open surgery such as scissors, forceps, hemostats, blade-handles and in laparoscopic surgery instruments such as graspers, cutter, needle holder and cautery. Further, it also used to make the rotating ball in the tips of ballpoint pens, bridal jewelry, and gauge blocks, which is useful for producing precision lengths in dimensional metrology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12070-12-1
195.85
UONOETXJSWQNOL-UHFFFAOYSA-N
methanidylidynetungstenylium
CW
MFCD00011464
tungsten carbide, tungsten iv carbide, rfdbds g@, tungsten carbide ds 60, methanidylidynetungstenylium, methanidylidynetungsten 1+, tungsten-carbide/cobalt co=8wt% powder nano
[C-]#[W+]

Specifications

2870°C
12070-12-1
100 g
CW
tungsten carbide, tungsten iv carbide, rfdbds g@, tungsten carbide ds 60, methanidylidynetungstenylium, methanidylidynetungsten 1+, tungsten-carbide/cobalt co=8wt% powder nano
UONOETXJSWQNOL-UHFFFAOYSA-N
methanidylidynetungstenylium
195.86
15.63
Gray
Powder
99.5% metal basis
MFCD00011464
Soluble in nitric acid and hydrofluroic acid. Insoluble in water.
[C-]#[W+]
195.85
Odorless
Tungsten carbide
Videos
Safety and Handling

Safety and Handling

P261-P271-P304+P340-P311-P501c

H331

EINECSNumber : 235-123-0

RTECSNumber : YO7250000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only