missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Tumor Necrosis Factor-alpha, human
$221.10
Specifications
| Molecular Weight (g/mol) | 17.5 kd |
|---|---|
| Packaging | Plastic bottle |
| Form | Lyophilized |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ67162EXE
|
Thermo Scientific Chemicals
J67162EXE |
10μg |
Each for $221.10
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 17.5 kd | |
| Plastic bottle | |
| Lyophilized |
RUO – Research Use Only