missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tubulin Tracker™ Variety Pack

Catalog No. T34079
Change view
Click to view available options
Quantity:
2 x 60 Slides
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34079 2 x 60 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T34079 Supplier Invitrogen™ Supplier No. T34079

The Tubulin Tracker Variety Pack is a kit containing Tubulin Tracker Green and Tubulin Tracker Deep Red, which provide green and deep-red fluorescence, respectively, when bound to polymerized tubulin in live cells.

Tubulin Tracker Green provides fluorescence with excitation maxima of 494 nm and emission maxima of 522 nm and can be detected with standard FITC/GFP filter settings using a fluorescence microscope. Tubulin Tracker Deep Red provides fluorescence with excitation maxima of 652 nm and emission maxima of 669 nm and can be detected using standard Cy5 filter settings.

Specifications

Content And Storage Contains sufficient material for at least 60 slides each for Tubulin Tracker Green and Tubulin Tracker Deep Red at 1 mL / slide and 1 x working concentration.This kit contains one Tubulin Tracker Green kit (Cat. No. T34078) and one Tubulin Tracker
Excitation Wavelength Range Tubulin Tracker Deep Red: 652/669, Tubulin Tracker Green: 494/522
Quantity 2 x 60 Slides
Product Line TubulinTracker
Shipping Condition Approved for shipment on Wet or Dry Ice
Label Type UV-Vis Labels
Product Type Tubulin Tracker
Sub Cellular Localization Tubulin
What is the difference between Tubulin Tracker Green (Cat No. T34078) and Tubulin Tracker Green, for live cell imaging (Cat. No. T34075)?

Both products include the same Tubulin Tracker Green reagent. The difference is in the packaging and amount of Tubulin Tracker Green reagent contained. Tubulin Tracker Green, for live cell imaging (Cat. No. T34075) includes enough Tubulin Tracker Green reagent for 300 slides whereas Tubulin Tracker Green (Cat No. T34078) contains enough Tubulin Tracker Green reagent for 60 slides. Also, Tubulin Tracker Green, for live cell imaging (Cat. No. T34075) does not include Probenecid.

With Tubulin Tracker Deep Red and Tubulin Tracker Green, can I fix samples after labeling?

No. The labeling is not retained upon fixation. Imaging of the samples should be done only on live cells. To limit photobleaching while viewing live cells, use ProLong Live Antifade Reagent, for live cell imaging (Cat No. P36974 and P36975).

How is Docetaxel in Tubulin Tracker Deep Red different from Taxol in Tubulin Tracker Green? Are there any differences in the binding or cytotoxicity between the two reagents?

Docetaxel has a slightly higher binding affinity than Taxol and exhibits longer retention time due to better uptake and slower efflux, but for use in labeling cultured cells, these differences should be minimal. More information comparing the properties of these two reagents is provided in this publication: https://www.ncbi.nlm.nih.gov/pubmed/1671606 https://www.cell.com/fulltext/S1074-5521(05)00301-7

Is Tubulin Tracker Deep Red acetylated? Does it require intracellular esterase activity to make the reagent active?

Tubulin Tracker Deep Red is not acetylated. It is inherently cell permeable and is not modified further by any cellular enzymatic activity to fluoresce or bind to actin.

Do I need to use Probenecid when using Tubulin Tracker Deep Red?

The use of Probenecid is optional. Its requirement is dependent upon the cell type. Some cell types actively efflux the reagent out of the cells within a short time, limiting detection, other cell types do not exhibit this activity or exhibit very limited activity.

For Research Use Only. Not for use in diagnostic procedures.