missing translation for 'onlineSavingsMsg'
Learn More

Tubulin α, detyrosinated Mouse anti-Human, Clone: AA12, MilliporeSigma™

Catalog No. MAB5566I25
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5566I25 25 μL
MAB5566I100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAB5566I25 Supplier MilliporeSigma Supplier No. MAB5566I25UL
Only null left

Mouse Monoclonal Antibody

Tubulin α, detyrosinated Monoclonal antibody specifically detects Tubulin α, detyrosinated in Human samples. It is validated for Western Blotting.

Specifications

Antigen Tubulin α, detyrosinated
Applications Western Blot
Classification Monoclonal
Clone AA12
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen A linear peptide corresponding to 6 amino acids (GEEEGE) from the C-terminal region of human Tubulin a.
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG2a
Show More Show Less