missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Tubular Push/Pull Balances

Tubular Push/Pull Balances are ideal tools for studying mass (using grams) and force (using newtons) in a visually and kinesthetically effective manner.

$30.21 - $33.46

Specifications

Product Type Pulley
For Use With (Equipment) Weights
Material Plastic
View More Specs

Products 5
Catalog Number Mfr. No. Capacity (English) Color Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Color Price Quantity & Availability  
S88326
View Documents Promotion Details
Eisco™
PH0035A
2.5 N Blue
Each for $30.21
Only null left
 
S88327
View Documents Promotion Details
Eisco™
PH0035B
Subdivision: 17.6 oz Green
Each for $30.21
Only null left
 
S88328
View Documents Promotion Details
Eisco™
PH0035D
20 N Red
Each for $30.21
Only null left
 
S88329
View Documents Promotion Details
Eisco™
PH0035E
Subdivision: 6.6 lb White
Each for $30.21
Only null left
 
S88330
View Documents Promotion Details
Eisco™
PH0035G
Subdivision: 11 lb Yellow
Each for $33.46
Only null left
 
Description

Description

  • High quality push balance
  • Transparent tubular body with calibrated dual scales
  • Use grams (g) or kilograms (kg) for mass activities
  • Use Newtons (N) for force activities

Warranty

1 year limited manufacturer warranty

Specifications

Specifications

Pulley
Weights
Plastic
Videos
SDS
Documents

Documents