missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TUBG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570098
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570098 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570098 Supplier Invitrogen™ Supplier No. PA570098
Only null left

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 85%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Yeast: 83%; Zebrafish: 83%.

Tubulin is the major constituent of microtubules. The gamma chain is found at microtubule organizing centers (MTOC) such as the spindle poles or the centrosome. Pericentriolar matrix component that regulates alpha/beta chain minus-end nucleation, centrosome duplication and spindle formation.

Specifications

Antigen TUBG2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Tubg2
Gene Accession No. Q9NRH3
Gene Alias AI504772; Gamma-2-tubulin; Tubg2; Tubgl; tubulin gamma 2; tubulin gamma-2 chain; tubulin, gamma 2; Unknown (protein for MGC:133882)
Gene Symbols Tubg2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human TUBG2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27175
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less