missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TUBA1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143782
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143782 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143782 Supplier Invitrogen™ Supplier No. PA5143782

Rabbit Polyclonal Antibody

The antibody detects a 50 kDa* protein corresponding to the molecular mass of alpha-Tubulin on SDS-PAGE immunoblots of human smooth muscle cells (A7r5) and mouse brain, as well as human recombinant a1-tubulin.

TUBA1C (Tubulin Alpha 1c) is a Protein Coding gene. Diseases associated with TUBA1C include Distal Hereditary Motor Neuropathy, Type Ii. Among its related pathways are Development Slit-Robo signaling and Gap junction. Gene Ontology (GO) annotations related to this gene include GTP binding and structural molecule activity. An important paralog of this gene is TUBA1B.

Specifications

Antigen TUBA1C
Applications Western Blot
Classification Polyclonal
Concentration ~1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene TUBA1C
Gene Accession No. P68373, Q6AYZ1, Q9BQE3
Gene Alias alpha-tubulin 6; Alpha-tubulin isotype M-alpha-6; bcm948; Detyrosinated tubulin alpha-1C chain; M[a]6; Tuba1c; TUBA6; tubulin alpha 1c; tubulin alpha-1C chain; Tubulin alpha-6 chain; tubulin, alpha 1C; tubulin, alpha 6
Gene Symbols TUBA1C
Host Species Rabbit
Immunogen alpha6-Tubulin synthetic peptide (coupled to KLH) corresponds to amino acid residues from the central region of human alpha6-Tubulin. This sequence is well conserved in most alpha-tubulin isoforms from most eukaryotic species, but is not conserved in beta-Tubulin.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22146, 300218, 84790
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less