missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TTLL9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559719
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559719 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559719 Supplier Invitrogen™ Supplier No. PA559719

Rabbit Polyclonal Antibody

Immunogen sequence: GITWIMKPVA RSQGKGIFLF RRLKDIVDWR KDTRSSDDQK DDIPVENYVA QRYIE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 66%.

Specifications

Antigen TTLL9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTLL9
Gene Accession No. Q3SXZ7
Gene Alias C20orf125; dJ310O13.1; Probable tubulin polyglutamylase TTLL9; TTLL9; tubulin tyrosine ligase like 9; tubulin tyrosine ligase-like family, member 9; tubulin--tyrosine ligase-like protein 9
Gene Symbols TTLL9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTLL9. Recombinant protein control fragment (Product #RP-99425).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 164395
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less