Learn More
Thermo Scientific™ TSV Series 72 cu.ft. Lab Refrigerator
Thermo Scientific TSV Series 72 cu.ft. lab refrigerators offer a convenient option for storage of non-critical samples and reagents. Featuring three self-closing glass doors, the 72 cu. ft. TSV Series Lab Refrigerators offer an economical solution for users who need significant space for temperature controlled storage.
Supplier: Thermo Scientific™ TSV72RSGA
Description
Thermo Scientific TSV Series 72 cu.ft. lab refrigerators offer a convenient option for storage of non-critical samples and reagents. Featuring three self-closing glass doors, the 72 cu. ft. TSV Series Lab Refrigerators offer an economical solution for users who need significant space for temperature controlled storage. Additionally, TSV Series 72 cu.ft. Lab Refrigerators come with remote alarm contacts, high and low temperature alarms, and an adjustbale setpoint with a digital controller.
- Three self-closing glass doors
- Remote alarm contacts
- High and low temperature alarms
- Digital controller with adjustable temperature setpoint
- Door ajar alarm
- Optional stainless steel interior
Specifications
| 72 cu. ft. | |
| 4°C to 6°C | |
| R290 | |
| 3 | |
| 100 lb. | |
| Stainless Steel | |
| cULus | |
| Digital | |
| 1 rear access port | |
| Automatic | |
| Key lock | |
| 75.7 x 32 x 81 in. (1924 x 813 x 2057 mm) | |
| 3 | |
| 730 lb. |
| 2038 L | |
| 5°C | |
| Glass Self-closing with Lock | |
| 12 Adjustable | |
| 45 kg | |
| Stainless Steel | |
| 318 kg | |
| Yes, customer installed | |
| 2 years parts & labor, 3 additional years compressor parts | |
| SMART VUE Compatible, Remote Alarm Contacts | |
| Bottom-mounted | |
| 72 x 28.5 x 61.75 in. (1829 x 724 x 1569 mm) | |
| 120 V/60 Hz | |
| 318 kg |