Learn More
Thermo Scientific™ TSV Series 72 cu.ft. Lab Refrigerator

Thermo Scientific TSV Series 72 cu.ft. lab refrigerators offer a convenient option for storage of non-critical samples and reagents. Featuring three self-closing glass doors, the 72 cu. ft. TSV Series Lab Refrigerators offer an economical solution for users who need significant space for temperature controlled storage.
$30,601.71 - $36,948.73
Specifications
| Capacity (English) | 72 cu. ft. |
|---|---|
| Capacity (Metric) | 2038 L |
| Temperature Range | 4°C to 6°C |
| Temperature (Setpoint) | 5°C |
| Refrigerant | R290 |
| Catalog Number | Mfr. No. | Cabinet Material | Exterior Finish | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Cabinet Material | Exterior Finish | Price | Quantity & Availability | ||||
TSV72RPGA
![]() |
Thermo Scientific™
TSV72RPGA |
Painted Steel | Painted Steel |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
TSV72RSGA
![]() |
Thermo Scientific™
TSV72RSGA |
Stainless Steel | Stainless Steel |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Thermo Scientific TSV Series 72 cu.ft. lab refrigerators offer a convenient option for storage of non-critical samples and reagents. Featuring three self-closing glass doors, the 72 cu. ft. TSV Series Lab Refrigerators offer an economical solution for users who need significant space for temperature controlled storage. Additionally, TSV Series 72 cu.ft. Lab Refrigerators come with remote alarm contacts, high and low temperature alarms, and an adjustbale setpoint with a digital controller.
- Three self-closing glass doors
- Remote alarm contacts
- High and low temperature alarms
- Digital controller with adjustable temperature setpoint
- Door ajar alarm
- Optional stainless steel interior
Specifications
| 72 cu. ft. | |
| 4°C to 6°C | |
| R290 | |
| 3 | |
| 100 lb. | |
| cULus | |
| Yes, customer installed | |
| Automatic | |
| Key lock | |
| 75.7 x 32 x 81 in. (1924 x 813 x 2057 mm) | |
| ENERGY STAR | |
| 318 kg |
| 2038 L | |
| 5°C | |
| Glass Self-closing with Lock | |
| 12 Adjustable | |
| 45 kg | |
| Digital | |
| 2 years parts & labor, 3 additional years compressor parts | |
| SMART VUE Compatible, Remote Alarm Contacts | |
| Bottom-mounted | |
| 72 x 28.5 x 61.75 in. (1829 x 724 x 1569 mm) | |
| 730 lb. |