missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, B.R. 35 DEG -60°C, ACS, Spectrum™ Chemical

Catalog No. 18612198
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612198 20 L Steel Can
18612194 100 mL Amber Glass Bottle
18612195 500 mL Amber Glass Bottle
18612196 1 L Amber Glass Bottle
18612197 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612198 Supplier Spectrum Chemical Mfg Cor Supplier No. P104520LTMT
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 8032-32-4

Specifications

Acidity To Pass Test
CAS Max % 1
Quantity 20 L
Packaging Steel Can
Residue after Evaporation 1e-05
Boiling Point 20°C to 40°C
Grade Reagent
Chemical Name or Material Petroleum Ether

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.