missing translation for 'onlineSavingsMsg'
Learn More

Wanzl TSS Chrome Wire Basket

Chrome Wire Basket for TSS Shelving System.

$162.06 - $383.77

Specifications

For Use With (Application) Shelving
Includes Wire Basket
Color Chrome
Material Steel
View More Specs

Products 8
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
11222108
View Documents
Wanzl
BSK1824CH
18 x 24 in.
Each for $162.06
Only null left
 
11222109
View Documents
Wanzl
BSK1836CH
18 x 36 in.
Each for $198.06
Only null left
 
11222110
View Documents
Wanzl
BSK1848CH
18 x 48 in.
Each for $258.85
Only null left
 
11222111
View Documents
Wanzl
BSK1860CH
18 x 60 in.
Each for $323.00
Only null left
 
11222112
View Documents
Wanzl
BSK2424CH
24 x 24 in.
Each for $237.47
Only null left
 
11222113
View Documents
Wanzl
BSK2436CH
24 x 36 in.
Each for $257.72
Only null left
 
11222114
View Documents
Wanzl
BSK2448CH
24 x 48 in.
Each for $317.36
Only null left
 
11222115
View Documents
Wanzl
BSK2460CH
24 x 60 in.
Each for $383.77
Only null left
 
Description

Description

  • Add-on option for additional customization
  • Offered in our Chrome Plus (CH) finish. The heavy nickel plate process features a hard chromium finish with special brighteners to enhance and increase the life of the shelving.
Specifications

Specifications

Shelving
Wire Basket
Chrome
Steel
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'