missing translation for 'onlineSavingsMsg'
Learn More

Wanzl TSS Chrome Wire Basket

Catalog No. 11222115
Change view
Click to view available options
Dimensions (L x W):
18 x 24 in.
18 x 36 in.
18 x 48 in.
18 x 60 in.
24 x 24 in.
24 x 48 in.
24 x 60 in.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
11222115 24 x 60 in.
11222108 18 x 24 in.
11222109 18 x 36 in.
11222110 18 x 48 in.
11222111 18 x 60 in.
11222112 24 x 24 in.
11222114 24 x 48 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 11222115 Supplier Wanzl Supplier No. BSK2460CH
Only null left

Chrome Wire Basket for TSS Shelving System.

  • Add-on option for additional customization
  • Offered in our Chrome Plus (CH) finish. The heavy nickel plate process features a hard chromium finish with special brighteners to enhance and increase the life of the shelving.

Specifications

Dimensions (L x W) 24 x 60 in.
Includes Wire Basket
Width (English) 60 in.
Color Chrome
For Use With (Application) Shelving
Material Steel