missing translation for 'onlineSavingsMsg'
Learn More

Wanzl TSS Chrome Post

Chrome Plus Post for Shelving.

$26.96 - $31.62

Specifications

Color Chrome
Material Steel
For Use With (Application) Shelving
Includes Post
View More Specs

Products 2
Catalog Number Mfr. No. Width (English) Price Quantity  
Catalog Number Mfr. No. Width (English) Price Quantity  
11222048
View Documents
Wanzl
TP63CH
63 in.
Each for $26.96
Only null left
 
11222049
View Documents
Wanzl
TP74CH
74 in.
Each for $31.62
Only null left
 
Description

Description

  • Add-on option for additional customization
  • Special post lengths are available. Posts can also be joined together with a TRBCON for lengths higher than 86 in.
  • Posts higher than 86 in. should not be used with shelves less than 24 in. deep.
Specifications

Specifications

Chrome
Steel
Shelving
Post
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'