missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN6 Polyclonal Antibody, Invitrogen™
Description
Immunogen sequence: NSFKNNYEKA LKQYNSTGDY RSHAVDKIQN TLHCCGVTDY RDWTDTNYYS EKGFPKSCCK LEDCTPQRDA DKVNNEG
TSPAN6 Polyclonal antibody specifically detects TSPAN6 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Specifications
Specifications
| Antigen | TSPAN6 |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TSPAN6 |
| Gene Accession No. | O43657 |
| Gene Alias | 6720473L21Rik; A15 homolog; AI316786; Putative NF-kappa-B-activating protein 321; T245; T245 protein; tetraspan TM4SF; tetraspanin 6; tetraspanin TM4-D; Tetraspanin-6; Tm4sf; TM4SF6; Transmembrane 4 superfamily member 6; TSPAN6; Tspan-6; UNQ767/PRO1560 |
| Gene Symbols | TSPAN6 |
| Show More |