missing translation for 'onlineSavingsMsg'
Learn More

TSHZ2 Antibody, ProSci

Catalog No. 16594254
Change view
Click to view available options
Quantity:
20 μg Trial Size
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16594254 20 μg Trial Size
16919930 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16594254 Supplier ProSci Llc Supplier No. 67570020

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Specifications

Antigen TSHZ2
Applications ELISA, Western Blot, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution TSHZ2 antibody can be used for detection of TSHZ2 by Western blot at 1 μg/mL. Antibody can also be used for immunofluorescence starting at 20 μg/mL. For immunofluorescence start at 5 μg/mL. Antibody validated: Western Blot in mouse samples and Immunofluorescence in mouse samples. All other applications and species not yet tested.
Formulation TSHZ2 Antibody is supplied in PBS containing 0.02% sodium azide.
Gene Accession No. NP_775756
Gene Alias TSHZ2 Antibody: TSH2, ZABC2, ZNF218, OVC10-2, C20orf17, TSH2, Teashirt homolog 2, Ovarian cancer-related protein 10-2
Host Species Rabbit
Purification Method TSHZ2 Antibody is affinity chromatography purified via peptide column.
Quantity 20 μg Trial Size
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128553
Target Species Human, Mouse, Rat
Content And Storage TSHZ2 antibody can be stored at 4°C for three months and -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Product Type Antibody
Form Liquid
Isotype IgG
Research Category Stem Cell
Show More Show Less