missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TSFM Recombinant Rabbit Monoclonal Antibody (23GB4710)

Catalog No. MA553793
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553793 20 μL
MA553792 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553793 Supplier Invitrogen™ Supplier No. MA553793
Only null left

Rabbit Recombinant Monoclonal Antibody

TSFM Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Synthesis of the 13 mitochondrial-encoded proteins occurs on a dedicated mitochondrial translation apparatus similar to that found in prokaryotes and requires, in addition to the tRNAs and rRNAs encoded in mtDNA, the concerted action of several translation factors and a large number of mitochondrial ribosomal proteins, all of which are encoded by nuclear genes. The TSFM gene encodes a mitochondrial translation elongation factor (Smeitink et al., 2006 ).

Specifications

Antigen TSFM
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4710
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TSFM
Gene Accession No. P43897, Q9CZR8, Q9QYU2
Gene Alias 2310050B20Rik; 2A3-2; 9430024O13Rik; EFTS; EF-Ts; EF-Ts {ECO:0000255; EFTSMT; EF-TsMt; EF-TsMt {ECO:0000255; elongation factor Ts; elongation factor Ts, mitochondrial; elongation factor Ts, mitochondrial {ECO:0000255; HAMAP-Rule:MF_03135}; mitochondrial elongation factor Ts; translation elongation factor; Ts translation elongation factor, mitochondrial; TSFM; zgc:158429
Gene Symbols TSFM
Host Species Rabbit
Immunogen A synthesized peptide derived from human TSFM (40-66AA).
Purification Method Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10102, 66399, 679068
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less