missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Trypsin (1:250), powder

Catalog No. 27250018
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27250018 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 27250018 Supplier Gibco™ Supplier No. 27250018

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Trypsin (1:250), powder

Gibco™ Trypsin 1:250 powder is an irradiated mixture of proteases derived from porcine pancreas. Due to its digestive strength it is widely used for cell dissociation, during routine cell culture passaging, and primary tissue dissociation. We offer a variety of Gibco™ Trypsin and animal origin free TrypLE™ that feature:

  • Quality Testing
  • Documented Traceability
  • Dual-site cGMP Manufacturing

    Quality Testing
    Gibco™ Trypsin 1:250 is tested for endotoxin, mycoplasma, bacterial, fungal and viral contamination. Other quality tests include multiple activity assays, ash analysis and moisture analysis.

    Documented Traceability
    We can provide detailed documentation to meet your regulatory needs. Gibco™ Trypsin information available includes lot traceability, animal origin certificates, lot analyses, irradiation certificates, a viral inactivation summary, and supply chain transparency.

    cGMP Manufacturing and Quality System
    For supply chain continuity, we repack Gibco™ trypsin at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements, are certified to ISO 13485, and are registered with the FDA as medical device manufacturers.
  • Specifications

    Chelators No EDTA
    Content And Storage Storage conditions: 2–8°C.
    Shipping conditions: Ambient.
    Shelf life: 24 months from date of manufacture.
    Cell Type Mammalian Cells
    Concentration 250 X
    Culture Type Adherent Cell Culture
    Form Powder
    Product Type Cell Culture Dissociation Reagent
    Without Additives No Phenol Red
    Endotoxin Level Low
    Format Securitainer Capped Vial
    Product Line Gibco
    Quantity 100 g
    Shipping Condition Room Temperature
    Source Porcine Pancreas
    Tested For In vitro Bioassay
    Show More Show Less

    For Research Use or Further Manufacturing. Not for use in diagnostic procedures.