missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TrxR2 Recombinant Rabbit Monoclonal Antibody (23GB4925)
GREENER_CHOICE

Catalog No. p-200056356
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553834 100 μL
MA553835 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553834 Supplier Invitrogen™ Supplier No. MA553834
Only null left

Rabbit Recombinant Monoclonal Antibody

TrxR2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

The mammalian thioredoxin reductases (TrxRs) are a family of selenocysteine-containing pyridine nucleotide-disulfide oxidoreductases. All the mammalian TrxRs are homologous to glutathione reductase with respect to primary structure including the conserved redox catalytic site (-Cys-Val-Asn-Val-Gly-Cys-) but distinctively with a C-terminal extension containing a catalytically active penultimate selenocysteine (SeCys) residue in the conserved sequence (-Gly-Cys-SeCys-Gly). TrxR is homodimeric protein in which each monomer includes an FAD prosthetic group, a NADPH binding site and a redox catalytic site. Electrons are transferred from NADPH via FAD and the active-site disulfide to C-terminal SeCys-containing redox center, which then reduces the substrate like thioredoxin. The members of TrxR family are 55-58 kilodalton in molecular size and composed of three isoforms including cytosolic TrxR1, mitochondrial TrxR2, and TrxR3, known as Trx and GSSG reductase (TGR). TrxR plays a key role in protection of cells against oxidative stress and redox-regulatory mechanism of transcription factors and various biological phenomena.

Specifications

Antigen TrxR2
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4925
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TXNRD2
Gene Accession No. Q9NNW7
Gene Alias AA118373; ESTM573010; KIAA1652; Selenoprotein Z; SelZ; TGR; thioredoxin reductase 2; thioredoxin reductase 2, mitochondrial; thioredoxin reductase 3; thioredoxin reductase beta; thioredoxin reductase TR3; TR; TR beta; TR3; TR-BETA; Trxr2; Trxrd2; TXNRD2
Gene Symbols TXNRD2
Host Species Rabbit
Immunogen A synthesized peptide derived from human TXNRD2 (50-100AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10587
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less