missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TruScan G3 Handheld Raman Analyzer

Catalog No. 17028249
Change view
Click to view available options
Display:
Digital
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display
17028249 Digital
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17028249 Supplier Thermo Scientific™ Supplier No. 8000479101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TruScan G3 Handheld Raman Analyzer is a rugged, handheld system for rapid material identification in a pharmaceutical manufacturing environment.

  • Continues the evolution of raw material ID testing for pharmaceutical and biotechnology manufacturing quality control processes that require speed, ease and efficiency to maintain continuous manufacturing
  • Samples are often brought to a quality control lab for testing, increasing costs and decreasing productivity
  • Enables non-technical operators to rapidly perform raw material ID testing without the need of a lab, bringing lab-quality performance and results to any setting, whether that be a warehouse or a loading dock

Specifications

Product Type Handheld Raman Spectrometer
Display Digital
Barcode Supported Symbologies Most linear and 2D standards
Battery Type Rechargeable internal lithium ion battery >4 hours operation
Certifications/Compliance FDA 1040, 21 CFR part 11, CE certification, Ph. Eur. 8.7
Collection Optics NA = 0.33, 18 mm working distance; 0.2 to 2.5 mm spot size
Connections Ethernet
Description TruScan G3 Handheld Raman Analyzer
Dimensions (L x W x H) 8.2 x 4.2 x 1.7 in. (20.8 x 10.7 x 4.3 cm)
Exposure Automatic modes (12 ms minimum)
Includes Fingerprint reader for easy log in
Laser Excitation Wavelength 785nm ± 0.5 (2 cm-1 line width, stability <0.1 cm-1)
Laser Output 250 mW ±25
Operating Systems Microsoft Windows 7,8,10, Internet Explorer 11, Edge 25; Google Chrome 51
Optional Accessories Vial holder, universal tablet holder, cuvette holder
Ports Up to 10 simultaneous ports
Power Supply DC Wall Adapter 100/240 V, 50/60 Hz
Spectral Range 250 to 2875 cm-1
Spectral Resolution 8 to 10.5 cm-1(FWHM) across range
Temperature Range (Metric) -20°C to 40°C
Weight (English) 2 lb.
Weight (Metric) 0.9 kg
Show More Show Less