missing translation for 'onlineSavingsMsg'
Learn More

TRPM2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501557567
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501557567 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501557567 Supplier Bethyl Laboratories Inc Supplier No. A300414A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

TRPM2 belongs to the ion transport protein family which includes Sodium, Potassium, Calcium ion channels. This family is characterized by proteins possessing 6 transmembrane helices in which the last two helices flank a loop which determines ion selectivity. TRPM2 is activated by oxidative stress and confers susceptibility to cell death. Further, TRPM2 is a calcium-permeable cation channel that is regulated by free intracellular ADP-ribose. Several alternatively spliced transcript variants of the TRPM2 gene have been described, but their full-length nature is not known.

Specifications

Antigen TRPM2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene TRPM2
Gene Accession No. O94759
Gene Alias Transient receptor potential melastatin family 2, Trpm2-predicted
Gene Symbols TRPM2
Host Species Rabbit
Immunogen Between 650 and 700
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7226
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less