missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Troxipide 98.0+%, TCI America™
Supplier: TCI America T29091G
Specifications
| Troxipide | |
| 180°C | |
| C15H22N2O4 | |
| 1 g | |
| YSIITVVESCNIPR-UHFFFAOYSA-N | |
| 3,4,5-trimethoxy-N-piperidin-3-ylbenzamide | |
| 5597 | |
| ≥98.0% (HPLC,T) |
| 30751-05-4 | |
| White | |
| MFCD00866876 | |
| 3,4,5-Trimethoxy-N-(3-piperidyl)benzamide | |
| COC1=CC(=CC(=C1OC)OC)C(=O)NC2CCCNC2 | |
| 294.351 | |
| 294.35 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : CV5792600
TSCA : No