missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trospium Chloride 98.0+%, TCI America™
Supplier: TCI America T33051G
Specifications
| Trospium Chloride | |
| 262°C | |
| C25H30ClNO3 | |
| 1 g | |
| C1CC[N+]2(C1)C3CCC2CC(C3)OC(=O)C(C4=CC=CC=C4)(C5=CC=CC=C5)O.[Cl-] | |
| 427.969 | |
| 427.97 | |
| Crystalline Powder |
| 10405-02-4 | |
| White | |
| MFCD00865254 | |
| RVCSYOQWLPPAOA-VROPFNGYSA-M | |
| [(1S,5S)-spiro[8-azoniabicyclo[3.2.1]octane-8,1'-azolidin-1-ium]-3-yl] 2-hydroxy-2,2-diphenylacetate;chloride | |
| 73415998 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 10405-02-4 | |
| 427.969 | |
| RVCSYOQWLPPAOA-VROPFNGYSA-M | |
| [(1S,5S)-spiro[8-azoniabicyclo[3.2.1]octane-8,1'-azolidin-1-ium]-3-yl] 2-hydroxy-2,2-diphenylacetate;chloride |
| C25H30ClNO3 | |
| MFCD00865254 | |
| 73415998 | |
| C1CC[N+]2(C1)C3CCC2CC(C3)OC(=O)C(C4=CC=CC=C4)(C5=CC=CC=C5)O.[Cl-] |
Safety and Handling
RTECSNumber : WH1424000
TSCA : No