missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tropolone Tosylate 99.0+%, TCI America™
Supplier: TCI America T157425G
Specifications
| Tropolone Tosylate | |
| 160°C | |
| C14H12O4S | |
| 25 g | |
| AWUMFQREGHLKJB-UHFFFAOYSA-N | |
| 7-oxocyclohepta-1,3,5-trien-1-yl 4-methylbenzene-1-sulfonate | |
| 562886 | |
| ≥99.0% (GC,T) |
| 38768-08-0 | |
| Yellow | |
| MFCD00671574 | |
| 2-Toluenesulfonyloxytropone, 2-Tosyloxytropone | |
| CC1=CC=C(C=C1)S(=O)(=O)OC1=CC=CC=CC1=O | |
| 276.31 | |
| 276.31 | |
| Crystalline Powder |
Chemical Identifiers
| 38768-08-0 | |
| 276.31 | |
| AWUMFQREGHLKJB-UHFFFAOYSA-N | |
| 562886 | |
| CC1=CC=C(C=C1)S(=O)(=O)OC1=CC=CC=CC1=O |
| C14H12O4S | |
| MFCD00671574 | |
| 2-Toluenesulfonyloxytropone, 2-Tosyloxytropone | |
| 7-oxocyclohepta-1,3,5-trien-1-yl 4-methylbenzene-1-sulfonate |
Safety and Handling
TSCA : No