missing translation for 'onlineSavingsMsg'
Learn More

Tropolone Tosylate 99.0+%, TCI America™

Catalog No. p-7136844
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15745G 5 g
T157425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15745G Supplier TCI America Supplier No. T15745G
Only null left

Chemical Identifiers

CAS 38768-08-0
Molecular Formula C14H12O4S
Molecular Weight (g/mol) 276.31
MDL Number MFCD00671574
InChI Key AWUMFQREGHLKJB-UHFFFAOYSA-N
Synonym 2-Toluenesulfonyloxytropone, 2-Tosyloxytropone
PubChem CID 562886
IUPAC Name 7-oxocyclohepta-1,3,5-trien-1-yl 4-methylbenzene-1-sulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC1=CC=CC=CC1=O

Specifications

Melting Point 160°C
Color Yellow
Quantity 5 g
Formula Weight 276.31
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tropolone Tosylate