missing translation for 'onlineSavingsMsg'
Learn More

Tropisetron Hydrochloride 98.0+%, TCI America™

Catalog No. T27435G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27435G 5 g
T27431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27435G Supplier TCI America Supplier No. T27435G

Chemical Identifiers

CAS 105826-92-4
Molecular Formula C17H21ClN2O2
Molecular Weight (g/mol) 320.817
MDL Number MFCD00210221
InChI Key XIEGSJAEZIGKSA-ICHLHHFLSA-N
Synonym sdz-ics 930
PubChem CID 71300439
IUPAC Name [(5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 1H-indole-3-carboxylate;hydrochloride
SMILES CN1C2CCC1CC(C2)OC(=O)C3=CNC4=CC=CC=C43.Cl

Specifications

Melting Point 285°C
Quantity 5 g
UN Number 2811
Formula Weight 320.82
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tropisetron Hydrochloride
RTECSNumber NL6012520
TSCA No
Recommended Storage Refrigerator