accessibility menu, dialog, popup

UserName

ClCH3N(R)(S)OOHNH

CAS RN 105826-92-4

IUPAC Name: hydrogen (1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl 1H-indole-3-carboxylate chloride
Synonyms: hydrogen tropisetron chloride
Molecular Weight (g/mol): 320.82
Molecular Formula: C17H21ClN2O2
InChi Key: XIEGSJAEZIGKSA-KOQCZNHOSA-N
SMILES: [H+].[Cl-].CN1[C@H]2CC[C@@H]1CC(C2)OC(=O)C1=CNC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results