missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Tropinone | |
| 43°C | |
| 218°C | |
| MFCD00005549 | |
| tropinone, 8-methyl-8-azabicyclo 3.2.1 octan-3-one, 3-tropanone, tropanone, tropanon, tropinon, tropionone, 3-tropinone, 8-azabicyclo 3.2.1 octan-3-one, 8-methyl, 1.alpha.h,5.alpha.h-tropan-3-one | |
| CN1C2CCC1CC(=O)C2 | |
| 139.20 | |
| 139.20 | |
| Crystalline Powder |
| 532-24-1 | |
| White-Yellow | |
| C8H13NO | |
| 5 g | |
| QQXLDOJGLXJCSE-UHFFFAOYNA-N | |
| 8-methyl-8-azabicyclo[3.2.1]octan-3-one | |
| 79038 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 532-24-1 | |
| 139.20 | |
| QQXLDOJGLXJCSE-UHFFFAOYNA-N | |
| 79038 | |
| CN1C2CCC1CC(=O)C2 |
| C8H13NO | |
| MFCD00005549 | |
| tropinone, 8-methyl-8-azabicyclo 3.2.1 octan-3-one, 3-tropanone, tropanone, tropanon, tropinon, tropionone, 3-tropinone, 8-azabicyclo 3.2.1 octan-3-one, 8-methyl, 1.alpha.h,5.alpha.h-tropan-3-one | |
| 8-methyl-8-azabicyclo[3.2.1]octan-3-one |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator