missing translation for 'onlineSavingsMsg'
Learn More

Tropinone 98.0+%, TCI America™

Catalog No. p-7136372
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T06265G 5 g
T062625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T06265G Supplier TCI America Supplier No. T06265G
Only null left

Chemical Identifiers

CAS 532-24-1
Molecular Formula C8H13NO
Molecular Weight (g/mol) 139.20
MDL Number MFCD00005549
InChI Key QQXLDOJGLXJCSE-UHFFFAOYNA-N
Synonym tropinone, 8-methyl-8-azabicyclo 3.2.1 octan-3-one, 3-tropanone, tropanone, tropanon, tropinon, tropionone, 3-tropinone, 8-azabicyclo 3.2.1 octan-3-one, 8-methyl, 1.alpha.h,5.alpha.h-tropan-3-one
PubChem CID 79038
IUPAC Name 8-methyl-8-azabicyclo[3.2.1]octan-3-one
SMILES CN1C2CCC1CC(=O)C2

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 218°C
Quantity 5 g
Formula Weight 139.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tropinone
TSCA Yes
Recommended Storage Refrigerator