missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Tropicamide USP
Description
Tropicamide USP is a white or practically white crystalline powder. It is odorless or having not more than a slight odor.
Specifications
Specifications
| Chemical Name or Material | Tropicamide USP |
| Packaging | Bottle, 60mL, Amber Glass, Wide Mouth Packer, 33-400 |
| CAS | 1508-75-4 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 284.357 |
| Quantity | 10 g |
| Percent Purity | 98 to 102 % |
| Grade | USP |