missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tromethamine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 77-86-1 |
| Physical Form | Neat |
| Linear Formula | C4H11 NO3 |
| MDL Number | MFCD00004679 |
| Synonym | Tromethamine; Tris |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Formula Weight | 121.14 |
| Grade | Certified Reference Material |