missing translation for 'onlineSavingsMsg'
Learn More

Trolamine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012322
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012322 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012322 Supplier Merck Emd Millipore Supplier No. PHR15785ML
Only null left
Pharmaceutical (small molecule)

Specifications

Packaging Ampule
Ignition Point 600°F
CAS 102-71-6
Vapor Pressure 0.01 mmHg (20°C)
Vapor Density 5.14 (vs. air)
Density 1.124 g/mL (at 25°C)
Flash Point 179°C - Closed Cup
Linear Formula (HOCH2 CH2)3 N
MDL Number MFCD00002855
Melting Point 17.9°C to 21°C
Boiling Point 190°C to 193°C (5 mmHg)
Synonym Triethanolamine; 2,2 ′,2 ′′-Nitrilotriethanol; Tris(2-hydroxyethyl)amine
Type Certified Reference Material
Quantity 5 mL
Formula Weight 149.19
Grade Certified Reference Material
Show More Show Less